Assay Details
Target Gene Details
Entrez Gene ID: | 151306 |
Gene Name: | G protein-coupled bile acid receptor 1 |
Gene Aliases: |
BG37, GPCR19, GPR131, M-BAR, TGR5 |
Location: |
Chr.2:218259496-218263861 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPBAR1 | NM_001077191.1 | 2 | 945 | NP_001070659.1 |
| NM_001077194.1 | 2 | 437 | NP_001070662.1 | |
| NM_001321950.1 | 2 | 811 | NP_001308879.1 | |
| NM_170699.2 | 2 | 324 | NP_733800.1 | |
| XM_011510743.1 | 2 | 1432 | XP_011509045.1 | |
| XM_017003467.1 | 3 | 1549 | XP_016858956.1 | |
| XM_017003468.1 | 3 | 2170 | XP_016858957.1 | |
| XM_017003469.1 | 3 | 1662 | XP_016858958.1 | |
| AB086170.1 | 1 | 77 | BAC65343.1 | |
| AB089307.1 | 1 | 77 | BAC55235.1 | |
| AK122658.1 | 2 | 811 | ||
| AK122660.1 | 2 | 945 | ||
| BC033625.1 | 2 | 308 | AAH33625.1 |
More Information
Set Membership: |
|

Genomic Map