Assay Details
Target Gene Details
Entrez Gene ID: | 51715 |
Gene Name: | RAB23, member RAS oncogene family |
Gene Aliases: |
HSPC137 |
Location: |
Chr.6:57186992-57222314 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB23 | NM_001278666.1 | 7 | 2495 | NP_001265595.1 |
| NM_001278667.1 | 7 | 2667 | NP_001265596.1 | |
| NM_001278668.1 | 7 | 2494 | NP_001265597.1 | |
| NM_016277.4 | 7 | 2964 | NP_057361.3 | |
| NM_183227.2 | 7 | 2518 | NP_899050.1 | |
| NR_103822.1 | 4 | 2169 | ||
| AF161486.1 | 7 | 2460 | AAF29101.1 | |
| CR749371.1 | 8 | 2698 | CAH18224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462949 | Chr.6:57109104 - 57234175 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map