Assay Details
Target Gene Details
Entrez Gene ID: | 84705 |
Gene Name: | GTP binding protein 3 (mitochondrial) |
Gene Aliases: |
COXPD23, GTPBG3, MSS1, MTGP1, THDF1 |
Location: |
Chr.19:17334982-17342731 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP3 | NM_001128855.2 | 1 | 55 | NP_001122327.1 |
| NM_001195422.1 | NP_001182351.1 | |||
| NM_032620.3 | 1 | 55 | NP_116009.2 | |
| NM_133644.3 | 1 | 55 | NP_598399.2 | |
| AF360742.1 | AAK39555.1 | |||
| AF360743.1 | AAK39556.1 | |||
| AF360744.1 | AAK39557.1 | |||
| AK027606.1 | BAB55228.1 | |||
| AK091035.1 | ||||
| AK291929.1 | ||||
| AK297953.1 | ||||
| AK310794.1 | ||||
| AK311442.1 | ||||
| AY078987.1 | AAL85492.1 | |||
| AY078988.1 | AAL85493.1 | |||
| BC017207.2 | AAH17207.1 | |||
| BC019261.1 | AAH19261.1 | |||
| BI856784.1 | ||||
| DA044241.1 | 1 | 55 | ||
| DA666199.1 | ||||
| DC339966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578741 | Chr.19:17335702 - 17339637 on Build GRCh38 | Gain |
|
| esv3643818 | Chr.19:17331902 - 17338827 on Build GRCh38 | Gain |
|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
| esv21604 | Chr.19:17332440 - 17339470 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map