Assay Details
Target Gene Details
Entrez Gene ID: | 80704 |
Gene Name: | solute carrier family 19 member 3 |
Gene Aliases: |
BBGD, THMD2, THTR2 |
Location: |
Chr.2:227685210-227718030 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC19A3 | NM_025243.3 | 6 | 2285 | NP_079519.1 |
| XM_005246874.3 | 7 | 2448 | XP_005246931.1 | |
| XM_011511931.2 | 6 | 2540 | XP_011510233.1 | |
| XM_011511932.1 | 6 | 2362 | XP_011510234.1 | |
| XM_011511933.1 | 6 | 2628 | XP_011510235.1 | |
| XM_017005030.1 | 7 | 2630 | XP_016860519.1 | |
| XM_017005031.1 | 7 | 2609 | XP_016860520.1 | |
| XM_017005032.1 | 7 | 2490 | XP_016860521.1 | |
| XM_017005033.1 | 7 | 2804 | XP_016860522.1 | |
| XM_017005034.1 | 7 | 2734 | XP_016860523.1 | |
| AF283317.1 | 6 | 2266 | AAK69539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000263 | Chr.2:227632997 - 227790216 on Build GRCh38 | Gain |
|
| nsv834561 | Chr.2:227550392 - 227703283 on Build GRCh38 | Gain |
|
| esv2721563 | Chr.2:227648287 - 227699345 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map