Assay Details
Target Gene Details
Entrez Gene ID: | 10999 |
Gene Name: | solute carrier family 27 member 4 |
Gene Aliases: |
ACSVL4, FATP4, IPS |
Location: |
Chr.9:128340517-128361470 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC27A4 | NM_005094.3 | 3 | 507 | NP_005085.2 |
| XM_017014222.1 | 4 | 380 | XP_016869711.1 | |
| AF055899.1 | 2 | 222 | AAD11623.1 | |
| AK027504.1 | 2 | 87 | ||
| AK290222.1 | 3 | 507 | ||
| AK314681.1 | 3 | 347 | ||
| BC009959.2 | AAH09959.1 | |||
| BC065003.1 | 3 | 506 | AAH65003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map