Assay Details
Target Gene Details
Entrez Gene ID: | 102724058 |
Gene Name: | uncharacterized LOC102724058 |
Gene Aliases: |
- |
Location: |
Chr.2:165957418-166036400 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724058 | NR_110598.1 | |||
| HY008183.1 | ||||
| HY011842.1 | ||||
| HY029566.1 |
Target Gene Details
Entrez Gene ID: | 6323 |
Gene Name: | sodium voltage-gated channel alpha subunit 1 |
Gene Aliases: |
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI |
Location: |
Chr.2:165989160-166149216 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN1A | NM_001165963.1 | 26 | 6268 | NP_001159435.1 |
| NM_001165964.1 | 26 | 6184 | NP_001159436.1 | |
| NM_001202435.1 | 28 | 6477 | NP_001189364.1 | |
| NM_006920.4 | 26 | 6235 | NP_008851.3 | |
| XM_011511602.2 | 27 | 6544 | XP_011509904.1 | |
| XM_011511605.2 | 28 | 6608 | XP_011509907.1 | |
| XM_011511606.2 | 28 | 6552 | XP_011509908.1 | |
| XM_017004644.1 | 29 | 6723 | XP_016860133.1 | |
| XM_017004650.1 | 27 | 6510 | XP_016860139.1 | |
| XM_017004651.1 | 28 | 6415 | XP_016860140.1 | |
| XM_017004652.1 | 27 | 6455 | XP_016860141.1 | |
| XM_017004653.1 | 28 | 6562 | XP_016860142.1 | |
| XM_017004654.1 | 28 | 6643 | XP_016860143.1 | |
| AF225985.1 | 26 | 6238 | AAK00217.1 | |
| AK094487.1 | ||||
| AK293759.1 | 2 | 1526 | ||
| AK294900.1 | 6 | 2011 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834445 | Chr.2:165800387 - 165995152 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113696479] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map