Assay Details
Target Gene Details
Entrez Gene ID: | 151354 |
Gene Name: | family with sequence similarity 84 member A |
Gene Aliases: |
NSE1, PP11517 |
Location: |
Chr.2:14632686-14640046 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM84A | NM_145175.2 | 2 | 407 | NP_660158.2 |
| AF370404.1 | 2 | 353 | AAQ15240.1 | |
| AJ417080.1 | 2 | 361 | CAD10038.1 | |
| AK092654.1 | 2 | 376 | BAC03938.1 | |
| AK314463.1 | 2 | 376 | ||
| BC026346.1 | 1 | 401 | AAH26346.1 | |
| BC052284.1 | 2 | 356 | AAH52284.1 | |
| BI547868.1 | 2 | 412 |

Genomic Map