Assay Details
Target Gene Details
Entrez Gene ID: | 80777 |
Gene Name: | cytochrome b5 type B |
Gene Aliases: |
CYB5-M, CYPB5M, OMB5 |
Location: |
Chr.16:69424595-69466264 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5B | NM_030579.2 | 5 | 2990 | NP_085056.2 |
| AK098148.1 | 1 | 1125 | ||
| AK125218.1 | 1 | 528 |
More Information
Set Membership: |
|

Genomic Map