Assay Details
Target Gene Details
Entrez Gene ID: | 1244 |
Gene Name: | ATP binding cassette subfamily C member 2 |
Gene Aliases: |
ABC30, CMOAT, DJS, MRP2, cMRP |
Location: |
Chr.10:99782598-99853741 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC2 | NM_000392.4 | NP_000383.1 | ||
| XM_006717631.3 | XP_006717694.1 | |||
| XM_011539291.2 | XP_011537593.1 | |||
| XM_017015675.1 | XP_016871164.1 | |||
| BC136419.1 | ||||
| CB120555.1 | ||||
| U49248.1 | AAB09422.1 | |||
| U63970.1 | AAB39892.1 | |||
| U66683.1 | ||||
| X96395.2 | CAA65259.2 |
More Information
Set Membership: |
|

Genomic Map