Assay Details
Target Gene Details
Entrez Gene ID: | 51720 |
Gene Name: | ubiquitin interaction motif containing 1 |
Gene Aliases: |
RAP80, X2HRIP110 |
Location: |
Chr.5:176905005-177022640 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016283 | Chr.5:176904803 - 176968410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map