Assay Details
Target Gene Details
Entrez Gene ID: | 200942 |
Gene Name: | kelch domain containing 8B |
Gene Aliases: |
CHL |
Location: |
Chr.3:49171120-49176487 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHDC8B | NM_173546.2 | 2 | 399 | NP_775817.1 |
| XM_005264938.2 | 2 | 864 | XP_005264995.1 | |
| XM_005264940.4 | XP_005264997.1 | |||
| XM_006713015.2 | 2 | 864 | XP_006713078.1 | |
| XM_006713016.2 | 2 | 864 | XP_006713079.1 | |
| AK294160.1 | 2 | 386 | ||
| AY129011.1 | 2 | 356 | AAM98754.1 | |
| BC039083.1 | 2 | 349 | AAH39083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008750 | Chr.3:49171382 - 49324968 on Build GRCh38 | Gain |
|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|

Genomic Map