Assay Details
Target Gene Details
Entrez Gene ID: | 221468 |
Gene Name: | transmembrane protein 217 |
Gene Aliases: |
C6orf128, dJ355M6.2 |
Location: |
Chr.6:37212178-37259034 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM217 | NM_001286401.1 | 3 | 1684 | NP_001273330.1 |
| NM_145316.3 | 4 | 1251 | NP_660359.2 | |
| NR_104442.1 | 3 | 1153 | ||
| NR_104443.1 | 2 | 1068 | ||
| XM_011514369.2 | 2 | 579 | XP_011512671.1 | |
| XM_011514370.2 | 2 | 569 | XP_011512672.1 | |
| AK098666.1 | 4 | 1210 | BAC05371.1 | |
| AW188099.1 | 1 | 343 | ||
| BC026012.1 | 3 | 1123 | AAH26012.1 | |
| BC029657.1 | 2 | 549 | AAH29657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527191 | Chr.6:37206017 - 37212747 on Build GRCh38 | Loss |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|

Genomic Map