Assay Details
Target Gene Details
Entrez Gene ID: | 79621 |
Gene Name: | ribonuclease H2 subunit B |
Gene Aliases: |
AGS2, DLEU8 |
Location: |
Chr.13:50909678-50970462 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH2B | NM_001142279.2 | NP_001135751.1 | ||
| NM_024570.3 | NP_078846.2 | |||
| XM_006719867.3 | XP_006719930.1 | |||
| XM_011535229.1 | XP_011533531.1 | |||
| XM_011535230.2 | XP_011533532.1 | |||
| XM_011535231.2 | XP_011533533.1 | |||
| XM_011535233.2 | XP_011533535.1 | |||
| XM_017020747.1 | XP_016876236.1 | |||
| AK021774.1 | BAB13892.1 | |||
| AK223340.1 | BAD97060.1 | |||
| AY764036.1 | AAX13343.1 | |||
| BC001397.2 | AAH01397.1 | |||
| BC005088.1 | ||||
| BC007332.2 | AAH07332.1 | |||
| BC010174.2 | AAH10174.1 | |||
| BC036744.1 | AAH36744.1 | |||
| BX382357.2 | ||||
| DB221997.1 | ||||
| DT216141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map