Assay Details
Target Gene Details
Entrez Gene ID: | 3850 |
Gene Name: | keratin 3 |
Gene Aliases: |
CK3, K3 |
Location: |
Chr.12:52789685-52796108 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT3 | NM_057088.2 | NP_476429.2 | ||
| AJ628418.1 | CAF31522.1 | |||
| AK314987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| esv3629524 | Chr.12:52688070 - 52835680 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map