Assay Details
Target Gene Details
Entrez Gene ID: | 28972 |
Gene Name: | signal peptidase complex subunit 1 |
Gene Aliases: |
HSPC033, SPC1, SPC12, YJR010C-A |
Location: |
Chr.3:52705841-52708182 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPCS1 | NM_014041.3 | NP_054760.3 | ||
| AF092138.1 | AAD40380.1 | |||
| AK027426.1 | ||||
| AX775783.1 | CAE11591.1 | |||
| AX775785.1 | CAE11592.1 | |||
| AX775787.1 | CAE11593.1 | |||
| AX775833.1 | CAE11616.1 | |||
| BC000884.1 | AAH00884.1 | |||
| BI826431.1 | ||||
| BQ423345.1 | ||||
| CA309210.1 | ||||
| L38852.1 | AAL31361.1 |
More Information
Set Membership: |
|

Genomic Map