Assay Details
Target Gene Details
Entrez Gene ID: | 6616 |
Gene Name: | synaptosome associated protein 25 |
Gene Aliases: |
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2 |
Location: |
Chr.20:10218694-10307420 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAP25 | NM_001322903.1 | NP_001309832.1 | ||
| NM_001322904.1 | NP_001309833.1 | |||
| NM_001322905.1 | NP_001309834.1 | |||
| NM_001322906.1 | NP_001309835.1 | |||
| NM_001322907.1 | NP_001309836.1 | |||
| NM_001322908.1 | NP_001309837.1 | |||
| NM_001322909.1 | NP_001309838.1 | |||
| NM_001322910.1 | NP_001309839.1 | |||
| NM_003081.4 | NP_003072.2 | |||
| NM_130811.3 | NP_570824.1 | |||
| XM_017028022.1 | XP_016883511.1 | |||
| XM_017028023.1 | 1 | 136 | XP_016883512.1 | |
| AK094560.1 | ||||
| AK098347.1 | ||||
| AK223617.1 | BAD97337.1 | |||
| BI552777.1 | ||||
| BI602062.1 | ||||
| BI604120.1 | ||||
| D21267.1 | BAA22370.1 | |||
| DA072367.1 | ||||
| DA162693.1 | 1 | 136 |
Target Gene Details
Entrez Gene ID: | 100131208 |
Gene Name: | SNAP25 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:10023812-10219506 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAP25-AS1 | NR_040710.1 | |||
| AK124961.1 | ||||
| DA318406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv952985 | Chr.20:10217253 - 10219452 on Build GRCh38 | Deletion |
|
| nsv833919 | Chr.20:10035753 - 10230987 on Build GRCh38 | Gain |
|
| esv2674016 | Chr.20:10216797 - 10220344 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map