Assay Details
Target Gene Details
Entrez Gene ID: | 117194 |
Gene Name: | MAS related GPR family member X2 |
Gene Aliases: |
MGRG3, MRGX2 |
Location: |
Chr.11:19054455-19060681 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRX2 | NM_001303615.1 | 3 | 1386 | NP_001290544.1 |
| NM_054030.3 | 2 | 1076 | NP_473371.1 | |
| BC063450.1 | 2 | 1076 | AAH63450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv975890 | Chr.11:19055386 - 19056079 on Build GRCh38 | Duplication |
|
| esv2759806 | Chr.11:18797182 - 19060521 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map