Assay Details
Target Gene Details
Entrez Gene ID: | 51804 |
Gene Name: | SIX homeobox 4 |
Gene Aliases: |
AREC3 |
Location: |
Chr.14:60709538-60724321 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIX4 | NM_017420.4 | 3 | 1986 | NP_059116.3 |
| XM_005267759.2 | 4 | 1965 | XP_005267816.1 | |
| AK308528.1 | 4 | 1343 | ||
| BC098135.1 | 3 | 1863 | AAH98135.1 | |
| BC098282.1 | 3 | 1863 | AAH98282.1 | |
| BC099722.1 | 3 | 1863 | AAH99722.1 | |
| BC101934.1 | 3 | 1863 | AAI01935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564880 | Chr.14:60397865 - 60916029 on Build GRCh38 | Gain |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map