Assay Details
Target Gene Details
Entrez Gene ID: | 10328 |
Gene Name: | ER membrane protein complex subunit 8 |
Gene Aliases: |
C16orf2, C16orf4, COX4NB, FAM158B, NOC4 |
Location: |
Chr.16:85778624-85799744 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC8 | NM_001142288.1 | 1 | 269 | NP_001135760.1 |
| NM_006067.4 | 1 | 269 | NP_006058.1 | |
| XM_011522813.2 | 1 | 471 | XP_011521115.1 | |
| AF005888.1 | 1 | 138 | AAB94489.1 | |
| AK313977.1 | 1 | 251 | ||
| BC001472.2 | 1 | 221 | AAH01472.1 | |
| BC005886.2 | 1 | 221 | AAH05886.1 | |
| BC007445.1 | 1 | 212 | AAH07445.1 | |
| BC020250.1 | 1 | 221 | AAH20250.1 | |
| BE855452.1 | 1 | 143 | ||
| BP307869.1 | 1 | 274 | ||
| BT007132.1 | AAP35796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833316 | Chr.16:85697332 - 85900033 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|

Genomic Map