Assay Details
Target Gene Details
Entrez Gene ID: | 340665 |
Gene Name: | cytochrome P450 family 26 subfamily C member 1 |
Gene Aliases: |
FFDD4 |
Location: |
Chr.10:93061264-93068697 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP26C1 | NM_183374.2 | 3 | 543 | NP_899230.2 |
| AK127234.1 | ||||
| AK131099.1 | 3 | 999 | BAC85149.1 | |
| AY356349.1 | 3 | 543 | AAQ55485.1 |
More Information
Set Membership: |
|

Genomic Map