Assay Details
Target Gene Details
Entrez Gene ID: | 9468 |
Gene Name: | phosphate cytidylyltransferase 1, choline, beta |
Gene Aliases: |
CCTB, CTB |
Location: |
Chr.X:24558087-24672862 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCYT1B | NM_001163264.1 | 8 | 5091 | NP_001156736.1 |
| NM_001163265.1 | 9 | 4823 | NP_001156737.1 | |
| NM_004845.4 | 8 | 5148 | NP_004836.2 | |
| XM_017029977.1 | 9 | 4973 | XP_016885466.1 | |
| AK022741.1 | 1 | 1594 | ||
| AK226121.1 | 8 | 5091 | ||
| BC045634.1 | 8 | 4906 | AAH45634.2 |
More Information
Set Membership: |
|

Genomic Map