Assay Details
Target Gene Details
Entrez Gene ID: | 10248 |
Gene Name: | POP7 homolog, ribonuclease P/MRP subunit |
Gene Aliases: |
0610037N12Rik, RPP2, RPP20 |
Location: |
Chr.7:100706053-100707500 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POP7 | NM_005837.2 | 2 | 757 | NP_005828.2 |
| AJ572668.1 | 1 | 262 | ||
| BC001430.2 | 2 | 667 | AAH01430.1 | |
| BI666854.1 | 2 | 760 | ||
| U94316.1 | 2 | 663 | AAC24113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607933 | Chr.7:100642568 - 100767990 on Build GRCh38 | Gain |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv8184 | Chr.7:100643114 - 100748322 on Build GRCh38 | Loss |
|
| nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map