Assay Details
Target Gene Details
Entrez Gene ID: | 54567 |
Gene Name: | delta like canonical Notch ligand 4 |
Gene Aliases: |
AOS6, hdelta2 |
Location: |
Chr.15:40929333-40939060 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLL4 | NM_019074.3 | 11 | 2762 | NP_061947.1 |
| AB036931.1 | 11 | 2702 | BAB16085.1 | |
| AF279305.1 | 10 | 2332 | AAF81912.1 | |
| BC106950.2 | 11 | 2562 | AAI06951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1504 | Chr.15:40936351 - 40969374 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map