Assay Details
Target Gene Details
Entrez Gene ID: | 26219 |
Gene Name: | olfactory receptor family 1 subfamily J member 4 |
Gene Aliases: |
HSHTPCRX01, HTPCRX01, OR9-21 |
Location: |
Chr.9:122519141-122520082 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1J4 | NM_001004452.1 | 1 | 819 | NP_001004452.1 |
| BC131606.1 | 1 | 818 |
More Information
Set Membership: |
|

Genomic Map