Assay Details
Target Gene Details
Entrez Gene ID: | 27189 |
Gene Name: | interleukin 17C |
Gene Aliases: |
CX2, IL-17C |
Location: |
Chr.16:88638593-88640474 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL17C | NM_013278.3 | 2 | 244 | NP_037410.1 |
| AF152099.1 | 2 | 244 | AAF28105.1 | |
| AY358471.1 | 2 | 244 | AAQ88835.1 | |
| BC069152.1 | 1 | 195 | AAH69152.1 | |
| BC101835.1 | 2 | 239 | AAI01836.1 | |
| BC112196.1 | 2 | 239 | AAI12197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1926 | Chr.16:88626290 - 88660544 on Build GRCh38 | Insertion |
|
| nsv833325 | Chr.16:88553738 - 88698194 on Build GRCh38 | Loss |
|
| nsv952070 | Chr.16:88257295 - 88780192 on Build GRCh38 | Deletion |
|
| nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map