Assay Details
Target Gene Details
Entrez Gene ID: | 387778 |
Gene Name: | speedy/RINGO cell cycle regulator family member C |
Gene Aliases: |
RINGOC, Ringo2 |
Location: |
Chr.11:65170236-65173666 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPDYC | NM_001008778.1 | NP_001008778.1 | ||
| XM_017017705.1 | XP_016873194.1 | |||
| XM_017017706.1 | XP_016873195.1 | |||
| XM_017017707.1 | XP_016873196.1 | |||
| XM_017017708.1 | XP_016873197.1 | |||
| XM_017017709.1 | XP_016873198.1 | |||
| XM_017017710.1 | XP_016873199.1 | |||
| AY820305.1 | AAW30396.1 | |||
| BC137244.1 | ||||
| BC137246.1 | ||||
| BC144548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509418 | Chr.11:65164531 - 65202781 on Build GRCh38 | Insertion |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map