Assay Details
Target Gene Details
Entrez Gene ID: | 54929 |
Gene Name: | transmembrane protein 161A |
Gene Aliases: |
AROS-29 |
Location: |
Chr.19:19119616-19138501 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM161A | NM_001256766.1 | 2 | 121 | NP_001243695.1 |
| NM_017814.2 | 2 | 121 | NP_060284.1 | |
| XM_017026916.1 | 2 | 92 | XP_016882405.1 | |
| AK000429.1 | 2 | 78 | BAA91159.1 | |
| AK096964.1 | 2 | 86 | ||
| AK225424.1 | 2 | 74 | ||
| AY358088.1 | 2 | 105 | AAQ88455.1 | |
| BC005210.1 | 2 | 81 | AAH05210.1 | |
| BC016478.2 | 1 | 45 | AAH16478.2 | |
| DA268563.1 | 2 | 121 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833779 | Chr.19:19052683 - 19202282 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map