Assay Details
Target Gene Details
Entrez Gene ID: | 10110 |
Gene Name: | SGK2, serine/threonine kinase 2 |
Gene Aliases: |
H-SGK2, dJ138B7.2 |
Location: |
Chr.20:43558995-43585633 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGK2 | NM_001199264.1 | NP_001186193.1 | ||
| NM_016276.3 | NP_057360.2 | |||
| NM_170693.2 | NP_733794.1 | |||
| AF169034.2 | AAF12757.2 | |||
| AF186470.1 | AAG17012.1 | |||
| AK289596.1 | ||||
| AK308502.1 | ||||
| AY987010.1 | AAX88805.1 | |||
| BC014037.2 | ||||
| BC065511.1 | AAH65511.1 | |||
| BT020098.1 | AAV38901.1 | |||
| DA244897.1 | ||||
| DB183229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map