Assay Details
Target Gene Details
Entrez Gene ID: | 28514 |
Gene Name: | delta like canonical Notch ligand 1 |
Gene Aliases: |
DELTA1, DL1, Delta |
Location: |
Chr.6:170282200-170291075 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLL1 | NM_005618.3 | 11 | 3005 | NP_005609.3 |
| XM_005266934.3 | 11 | 2874 | XP_005266991.1 | |
| AF196571.1 | 11 | 2853 | AAF05834.1 | |
| AY358892.1 | 11 | 2572 | AAQ89251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759497 | Chr.6:170272663 - 170700831 on Build GRCh38 | Gain+Loss |
|
| nsv605554 | Chr.6:170263653 - 170308522 on Build GRCh38 | Loss |
|
| nsv823936 | Chr.6:170096017 - 170306104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map