Assay Details
Target Gene Details
Entrez Gene ID: | 27101 |
Gene Name: | calcyclin binding protein |
Gene Aliases: |
GIG5, PNAS-107, S100A6BP, SIP |
Location: |
Chr.1:174999435-175012027 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACYBP | NM_001007214.1 | NP_001007215.1 | ||
| NM_014412.2 | 1 | 62 | NP_055227.1 | |
| AK093425.1 | ||||
| BQ432793.1 | ||||
| CA951015.1 | 1 | 62 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001000 | Chr.1:174975071 - 175009717 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map