Assay Details
Target Gene Details
Entrez Gene ID: | 64785 |
Gene Name: | GINS complex subunit 3 |
Gene Aliases: |
PSF3 |
Location: |
Chr.16:58392394-58406144 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS3 | NM_001126129.1 | 4 | 1852 | NP_001119601.1 |
| NM_001126130.1 | 2 | 1501 | NP_001119602.1 | |
| NM_022770.3 | 3 | 1735 | NP_073607.2 | |
| AK023974.1 | 2 | 1407 | BAB14747.1 | |
| AK127454.1 | 1 | 2664 | ||
| AL137379.1 | 1 | 1071 | ||
| BC005879.1 | 3 | 1610 | AAH05879.1 | |
| BC014437.1 | 3 | 1629 | AAH14437.1 |
More Information
Set Membership: |
|

Genomic Map