Assay Details
Target Gene Details
Entrez Gene ID: | 339965 |
Gene Name: | coiled-coil domain containing 158 |
Gene Aliases: |
- |
Location: |
Chr.4:76313039-76421920 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC158 | NM_001042784.1 | 11 | 1657 | NP_001036249.1 |
| XM_005262974.4 | 12 | 1784 | XP_005263031.1 | |
| XM_006714204.3 | 12 | 1784 | XP_006714267.1 | |
| XM_011531908.2 | 13 | 1888 | XP_011530210.1 | |
| XM_011531909.2 | 13 | 1904 | XP_011530211.1 | |
| XM_011531910.2 | 12 | 2365 | XP_011530212.1 | |
| XM_011531911.1 | 12 | 1748 | XP_011530213.1 | |
| XM_011531912.2 | 12 | 1644 | XP_011530214.1 | |
| XM_011531913.1 | 11 | 1672 | XP_011530215.1 | |
| XM_011531914.2 | 11 | 1741 | XP_011530216.1 | |
| XM_011531915.2 | 12 | 1785 | XP_011530217.1 | |
| XM_011531916.2 | 11 | 1566 | XP_011530218.1 | |
| XM_011531917.1 | 12 | 1609 | XP_011530219.1 | |
| XM_017008160.1 | 13 | 2398 | XP_016863649.1 | |
| XM_017008161.1 | 7 | 849 | XP_016863650.1 | |
| AK098636.1 | BAC05359.1 | |||
| AL832445.1 | ||||
| BC086869.1 | 7 | 1027 | AAH86869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759262 | Chr.4:76343443 - 76534252 on Build GRCh38 | Loss |
|
| nsv1161115 | Chr.4:76364680 - 76371648 on Build GRCh38 | Deletion |
|

Genomic Map