Assay Details
Target Gene Details
Entrez Gene ID: | 8048 |
Gene Name: | cysteine and glycine rich protein 3 |
Gene Aliases: |
CLP, CMD1M, CMH12, CRP3, LMO4, MLP |
Location: |
Chr.11:19182030-19210571 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRP3 | NM_003476.4 | NP_003467.1 | ||
| AF121260.1 | AAF28868.1 | |||
| AK313289.1 | ||||
| BC005900.2 | AAH05900.1 | |||
| BC024010.1 | AAH24010.1 | |||
| BC057221.1 | AAH57221.1 | |||
| JN898958.1 | ||||
| U20324.1 | AAA91104.1 | |||
| U49837.1 | AAA92571.1 | |||
| U72899.1 | AAD00183.1 |
More Information
Set Membership: |
|

Genomic Map