Assay Details
Target Gene Details
Entrez Gene ID: | 5983 |
Gene Name: | replication factor C subunit 3 |
Gene Aliases: |
RFC38 |
Location: |
Chr.13:33818049-33973946 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFC3 | NM_002915.3 | NP_002906.1 | ||
| NM_181558.2 | NP_853536.2 | |||
| XM_011535172.2 | XP_011533474.1 | |||
| XM_011535173.2 | XP_011533475.1 | |||
| XM_011535174.2 | XP_011533476.1 | |||
| XM_011535175.1 | XP_011533477.1 | |||
| XM_017020680.1 | XP_016876169.1 | |||
| XM_017020681.1 | XP_016876170.1 | |||
| XM_017020682.1 | XP_016876171.1 | |||
| XM_017020683.1 | XP_016876172.1 | |||
| AK222572.1 | BAD96292.1 | |||
| AK296527.1 | ||||
| AK312544.1 | ||||
| AL596753.1 | ||||
| BC000149.2 | AAH00149.1 | |||
| BT019668.1 | AAV38474.1 | |||
| CX786577.1 | ||||
| DA943219.1 | ||||
| L07541.1 | AAB07268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054003 | Chr.13:33788101 - 34091373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map