Assay Details
Target Gene Details
Entrez Gene ID: | 6478 |
Gene Name: | siah E3 ubiquitin protein ligase 2 |
Gene Aliases: |
hSiah2 |
Location: |
Chr.3:150741123-150763476 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAH2 | NM_005067.5 | 1 | 621 | NP_005058.3 |
| AK223565.1 | 1 | 270 | BAD97285.1 | |
| BC013082.2 | 1 | 323 | AAH13082.1 | |
| U76248.1 | 1 | 521 | AAC51908.1 | |
| Y15268.1 | CAA75557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25944 | Chr.3:150762781 - 150763506 on Build GRCh38 | Loss |
|
| dgv8650n54 | Chr.3:150760231 - 150763449 on Build GRCh38 | Gain |
|
| nsv592037 | Chr.3:150762808 - 150762910 on Build GRCh38 | Loss |
|
| nsv1074633 | Chr.3:150762512 - 150763913 on Build GRCh38 | Deletion |
|
| nsv592041 | Chr.3:150762859 - 150763346 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map