Assay Details
Target Gene Details
Entrez Gene ID: | 5600 |
Gene Name: | mitogen-activated protein kinase 11 |
Gene Aliases: |
P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta |
Location: |
Chr.22:50263713-50270393 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK11 | NM_002751.6 | 11 | 1058 | NP_002742.3 |
| NR_110887.1 | 11 | 1058 | ||
| XM_011530706.1 | 9 | 782 | XP_011529008.1 | |
| AF001008.1 | 11 | 957 | AAC51250.1 | |
| AF001174.1 | 11 | 963 | AAC51373.1 | |
| AF031135.1 | 11 | 972 | AAC12714.1 | |
| AK291845.1 | 11 | 1009 | ||
| AK308969.1 | 10 | 895 | ||
| BC027933.1 | 11 | 1015 | AAH27933.1 | |
| CR456514.1 | 11 | 959 | CAG30400.1 | |
| U53442.1 | 11 | 1000 | AAB05036.1 | |
| U92268.1 | 11 | 961 | AAB66313.1 | |
| Y14440.1 | 11 | 963 | CAA74792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|
| nsv1066496 | Chr.22:50226183 - 50292445 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map