Assay Details
Target Gene Details
Entrez Gene ID: | 8819 |
Gene Name: | Sin3A associated protein 30 |
Gene Aliases: |
- |
Location: |
Chr.4:173370942-173377532 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP30 | NM_003864.3 | 1 | 244 | NP_003855.1 |
| AF055993.1 | 1 | 244 | AAC33316.1 | |
| AW236579.1 | ||||
| BC016757.1 | 1 | 239 | AAH16757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1074240 | Chr.4:173370048 - 173371649 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map