Assay Details
Target Gene Details
Entrez Gene ID: | 346517 |
Gene Name: | olfactory receptor family 6 subfamily V member 1 |
Gene Aliases: |
GPR138 |
Location: |
Chr.7:143052341-143053282 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6V1 | NM_001001667.1 | NP_001001667.1 | ||
| BC137044.1 | 1 | 55 |
More Information
Additional Information:
For this assay, SNP(s) [rs80058368] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map