Assay Details
Target Gene Details
Entrez Gene ID: | 81876 |
Gene Name: | RAB1B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.11:66268526-66277492 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB1B | NM_030981.2 | NP_112243.1 | ||
| XM_017018378.1 | XP_016873867.1 | |||
| AF092437.1 | AAP97212.1 | |||
| AJ245875.1 | CAC21570.1 | |||
| AK292086.1 | ||||
| AK315333.1 | ||||
| AL136635.1 | CAB66570.1 | |||
| BC071169.1 | AAH71169.1 | |||
| BG717501.1 | ||||
| CR533462.1 | CAG38493.1 | |||
| U66621.1 | AAC51196.1 |
More Information
Set Membership: |
|

Genomic Map