Assay Details
Target Gene Details
Entrez Gene ID: | 54790 |
Gene Name: | tet methylcytosine dioxygenase 2 |
Gene Aliases: |
KIAA1546, MDS |
Location: |
Chr.4:105145875-105279803 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TET2 | NM_017628.4 | 1 | 59 | NP_060098.3 |
| BX640738.1 | 1 | 60 | CAE45851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830025 | Chr.4:105067719 - 105216789 on Build GRCh38 | Loss |
|
| nsv950373 | Chr.4:105127544 - 105184443 on Build GRCh38 | Duplication |
|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
| esv3601659 | Chr.4:105145497 - 105197242 on Build GRCh38 | Loss |
|
| esv3601657 | Chr.4:105012463 - 105162253 on Build GRCh38 | Loss |
|
| nsv595007 | Chr.4:104948391 - 105445052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map