Assay Details
Target Gene Details
Entrez Gene ID: | 84688 |
Gene Name: | chromosome 9 open reading frame 24 |
Gene Aliases: |
CBE1, NYD-SP22, SMRP1, bA573M23.4 |
Location: |
Chr.9:34379019-34400833 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf24 | NM_001252195.1 | NP_001239124.1 | ||
| NM_032596.3 | NP_115985.2 | |||
| NM_147168.1 | 1 | 148 | NP_671697.1 | |
| NM_147169.2 | 1 | 148 | NP_671698.1 | |
| XM_005251616.4 | XP_005251673.1 | |||
| XM_005251618.4 | XP_005251675.1 | |||
| XM_011518055.2 | XP_011516357.1 | |||
| AA338648.1 | 1 | 148 | ||
| AF367474.1 | 1 | 87 | AAK53410.1 | |
| AK058087.1 | BAB71657.1 | |||
| AK094751.1 | 1 | 137 | BAC04412.1 | |
| AK225981.1 | ||||
| AW961020.1 | 1 | 148 | ||
| BC022084.1 | AAH22084.1 | |||
| BC029484.1 | AAH29484.1 | |||
| BI561111.1 | ||||
| CB854584.1 | 1 | 117 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv831542 | Chr.9:34273805 - 34461547 on Build GRCh38 | Gain |
|
| nsv1015595 | Chr.9:34342674 - 34474981 on Build GRCh38 | Gain |
|
| nsv1028418 | Chr.9:34360053 - 34401766 on Build GRCh38 | Gain |
|
| nsv474954 | Chr.9:34381506 - 34381506 on Build GRCh38 | Novel Sequence Insertion |
|
| nsv1016251 | Chr.9:34336344 - 34408206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map