Assay Details
Target Gene Details
Entrez Gene ID: | 84107 |
Gene Name: | Zic family member 4 |
Gene Aliases: |
- |
Location: |
Chr.3:147386048-147406893 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZIC4 | NM_001168378.1 | 5 | 2977 | NP_001161850.1 |
| NM_001168379.1 | 5 | 2804 | NP_001161851.1 | |
| NM_001243256.1 | 4 | 2485 | NP_001230185.1 | |
| NM_032153.5 | 5 | 3103 | NP_115529.2 | |
| NR_033118.2 | 3 | 2414 | ||
| NR_033119.2 | 3 | 2360 | ||
| NR_040762.1 | 3 | 2261 | ||
| AK124600.1 | 1 | 3439 | ||
| AK127733.1 | 3 | 2376 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822285 | Chr.3:147381294 - 147428675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map