Assay Details
Target Gene Details
Entrez Gene ID: | 10279 |
Gene Name: | protease, serine 16 |
Gene Aliases: |
TSSP |
Location: |
Chr.6:27247701-27256620 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS16 | NM_005865.3 | 10 | 1287 | NP_005856.1 |
| XM_017010161.1 | 9 | 1209 | XP_016865650.1 | |
| XM_017010162.1 | XP_016865651.1 | |||
| XM_017010163.1 | 9 | 996 | XP_016865652.1 | |
| XM_017010164.1 | XP_016865653.1 | |||
| XM_017010165.1 | 4 | 538 | XP_016865654.1 | |
| AF052514.1 | 10 | 1284 | AAC33563.1 | |
| AK093590.1 | 1 | 1176 | ||
| AK097918.1 | 4 | 538 | ||
| AK098033.1 | 1 | 463 | ||
| AK126160.1 | 1 | 1829 | ||
| AK226091.1 | ||||
| AK303106.1 | 5 | 608 | ||
| AK314037.1 | 10 | 1287 |
More Information
Set Membership: |
|

Genomic Map