Assay Details
Target Gene Details
Entrez Gene ID: | 3958 |
Gene Name: | lectin, galactoside binding soluble 3 |
Gene Aliases: |
CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2 |
Location: |
Chr.14:55129217-55145430 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGALS3 | NM_001177388.1 | NP_001170859.1 | ||
| NM_002306.3 | NP_002297.2 | |||
| NR_003225.2 | ||||
| XM_011536759.2 | XP_011535061.2 | |||
| AB006780.1 | BAA22164.1 | |||
| AB209391.1 | BAD92628.1 | |||
| AF266280.1 | AAG44701.1 | |||
| AK314929.1 | ||||
| BC001120.2 | AAH01120.1 | |||
| BC053667.1 | AAH53667.1 | |||
| BC068068.1 | AAH68068.1 | |||
| BG332134.1 | ||||
| BU681070.1 | ||||
| BX161455.1 | CAD61918.1 | |||
| BX641090.1 | CAE46042.1 | |||
| CD518618.1 | ||||
| CR456897.1 | CAG33178.1 | |||
| CR542097.1 | CAG46894.1 | |||
| M35368.1 | AAA88086.1 | |||
| M36682.1 | AAA36163.1 | |||
| M57710.1 | AAA35607.1 | |||
| M64303.1 | ||||
| S59012.1 | AAB26229.1 |
More Information
Set Membership: |
|

Genomic Map