Assay Details
Target Gene Details
Entrez Gene ID: | 503497 |
Gene Name: | membrane spanning 4-domains A13 |
Gene Aliases: |
- |
Location: |
Chr.11:60515382-60543424 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A13 | NM_001012417.2 | 3 | 394 | NP_001012417.2 |
| NM_001100909.1 | 3 | 394 | NP_001094379.1 | |
| NM_001278320.1 | 3 | 394 | NP_001265249.1 | |
| AA431602.1 | ||||
| AY324188.1 | 3 | 394 | AAQ94112.1 | |
| AY324189.1 | 3 | 394 | AAQ94113.1 | |
| AY324190.1 | 3 | 394 | AAQ94114.1 | |
| BC140799.1 | 3 | 268 | ||
| BC171746.1 | 2 | 629 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv337 | Chr.11:60487120 - 60519417 on Build GRCh38 | Insertion |
|
| nsv555152 | Chr.11:60508688 - 60649888 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map