Assay Details
Target Gene Details
Entrez Gene ID: | 26958 |
Gene Name: | coatomer protein complex subunit gamma 2 |
Gene Aliases: |
2-COP, gamma-2-COP |
Location: |
Chr.7:130506238-130668758 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPG2 | NM_001290033.1 | NP_001276962.1 | ||
| NM_012133.5 | NP_036265.3 | |||
| AB047847.1 | BAB17658.1 | |||
| AF157833.1 | AAF14271.1 | |||
| AF207598.1 | AAF19433.1 | |||
| AK022934.1 | BAB14315.1 | |||
| AK024414.1 | ||||
| AK291409.1 | ||||
| BC017443.1 | AAH17443.2 | |||
| BC110796.1 | AAI10797.1 | |||
| BG470623.1 | ||||
| BG704011.1 | ||||
| BX353590.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831132 | Chr.7:130511850 - 130727326 on Build GRCh38 | Gain+Loss |
|
| nsv831131 | Chr.7:130468986 - 130656327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map