Assay Details
Target Gene Details
Entrez Gene ID: | 1478 |
Gene Name: | cleavage stimulation factor subunit 2 |
Gene Aliases: |
CstF-64 |
Location: |
Chr.X:100820359-100841520 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSTF2 | NM_001306206.1 | NP_001293135.1 | ||
| NM_001306209.1 | NP_001293138.1 | |||
| NM_001325.2 | NP_001316.1 | |||
| AK095684.1 | ||||
| AK300600.1 | ||||
| BC017712.1 | AAH17712.1 | |||
| BC033135.1 | AAH33135.1 | |||
| BT009778.1 | AAP88780.1 | |||
| BX384743.2 | ||||
| EU616679.1 | ||||
| M85085.1 | AAA35724.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510850 | Chr.X:100727533 - 100827717 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map