Assay Details
Target Gene Details
Entrez Gene ID: | 54386 |
Gene Name: | TERF2 interacting protein |
Gene Aliases: |
DRIP5, RAP1 |
Location: |
Chr.16:75647704-75657442 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERF2IP | NM_018975.3 | 3 | 1594 | NP_061848.2 |
| AF250393.1 | 3 | 1500 | AAQ14259.1 | |
| AF289599.1 | 3 | 1502 | AAL55783.1 | |
| AK000669.1 | 3 | 1586 | BAA91317.1 | |
| AK298880.1 | 3 | 1292 | ||
| AK307901.1 | 4 | 1571 | ||
| BC004465.1 | 3 | 1530 | AAH04465.1 | |
| BC005841.2 | 3 | 1530 | AAH05841.1 | |
| BC022428.1 | 3 | 1545 | AAH22428.1 | |
| BC078171.1 | 3 | 1507 | AAH78171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1881 | Chr.16:75652316 - 75685543 on Build GRCh38 | Insertion |
|
| nsv1065543 | Chr.16:75597408 - 75766272 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map