Assay Details
Target Gene Details
Entrez Gene ID: | 7511 |
Gene Name: | X-prolyl aminopeptidase 1 |
Gene Aliases: |
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1 |
Location: |
Chr.10:109864766-109923553 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
More Information
Set Membership: |
|

Genomic Map