Assay Details
Target Gene Details
Entrez Gene ID: | 9580 |
Gene Name: | SRY-box 13 |
Gene Aliases: |
ICA12, Sox-13 |
Location: |
Chr.1:204073096-204127743 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX13 | NM_005686.2 | 1 | 51 | NP_005677.2 |
| XM_005245623.2 | 1 | 73 | XP_005245680.1 | |
| AK304801.1 | 1 | 54 | ||
| BC106038.1 | 1 | 70 | AAI06039.1 | |
| BQ935858.1 | 1 | 42 |
More Information
Set Membership: |
|

Genomic Map